RIE156330839 Rieke Metals, Inc.

4-(4-Chloro-3-methylphenyl)-4-oxobutyric acid

  • MDL number MFCD06201697
  • Cas Number : 54557-91-4
  • Molecular Weight : 226.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7238B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7238B
Availability details for
Quantity
  • Availability
  • Add to Cart