RIE156330845 Rieke Metals, Inc.

4-(4-Methoxy-3-methylphenyl)-4-oxobutyric acid

  • MDL number MFCD07186514
  • Cas Number : 33446-14-9
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/7239b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7239b
Availability details for
Quantity
  • Availability
  • Add to Cart