RIE156330846 Rieke Metals, Inc.

5-(4-Methoxy-3-methylphenyl)-5-oxovaleric acid

  • MDL number MFCD09801808
  • Cas Number : 4642-30-2
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7239C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7239C
Availability details for
Quantity
  • Availability
  • Add to Cart