RIE156330849 Rieke Metals, Inc.

Ethyl-4-(4-cyanophenyl)-4-oxobutyrate

  • MDL number MFCD01320301
  • Cas Number : 25370-40-5
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7571B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7571B
Availability details for
Quantity
  • Availability
  • Add to Cart