RIE156330852 Rieke Metals, Inc.

4-(2,3-Dichlorophenyl)-4-oxobutyric acid

  • MDL number MFCD02260926
  • Cas Number : 32003-41-1
  • Molecular Weight : 247.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7142B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7142B
Availability details for
Quantity
  • Availability
  • Add to Cart