RIE156330853 Rieke Metals, Inc.

3-Methyl-5-(3-methylphenyl)-5-oxovaleric acid

  • MDL number MFCD06201662
  • Cas Number : 845781-36-4
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7244

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7244
Availability details for
Quantity
  • Availability
  • Add to Cart