RIE156331102 Rieke Metals, Inc.

Ethyl 5-(2,4,5-Trimethylphenyl)-5-oxovalerate

  • MDL number MFCD09801978
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7725C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7725C
Availability details for
Quantity
  • Availability
  • Add to Cart