RIE156331105 Rieke Metals, Inc.

Ethyl 4-(2,3,5,6-Tetramethylphenyl)-4-oxobutanoate

  • MDL number MFCD09801981
  • Cas Number : 40888-48-0
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7727B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7727B
Availability details for
Quantity
  • Availability
  • Add to Cart