RIE156331115 Rieke Metals, Inc.

Ethyl 4-(5-Fluoro-2-methoxyphenyl)-4-oxobutanoate

  • MDL number MFCD09801991
  • Molecular Weight : 254.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7732B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7732B
Availability details for
Quantity
  • Availability
  • Add to Cart