RIE156331116 Rieke Metals, Inc.

Ethyl 5-(5-Fluoro-2-methoxyphenyl)-5-oxovalerate

  • MDL number MFCD09801992
  • Molecular Weight : 268.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7732C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7732C
Availability details for
Quantity
  • Availability
  • Add to Cart