RIE156331118 Rieke Metals, Inc.

Ethyl 5-(2-Methoxy-5-methylphenyl)-5-oxovalerate

  • MDL number MFCD09801994
  • Cas Number : 82054-02-2
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7733C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7733C
Availability details for
Quantity
  • Availability
  • Add to Cart