RIE156331120 Rieke Metals, Inc.

Ethyl 5-(3-Chloro-5-fluorophenyl)-5-oxovalerate

  • MDL number MFCD07782900
  • Molecular Weight : 272.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7734C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7734C
Availability details for
Quantity
  • Availability
  • Add to Cart