RIE156331126 Rieke Metals, Inc.

Ethyl 5-(4-Chloro-3-fluorophenyl)-5-oxovalerate

  • MDL number MFCD07782902
  • Molecular Weight : 272.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7737C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7737C
Availability details for
Quantity
  • Availability
  • Add to Cart