RIE156331129 Rieke Metals, Inc.

Ethyl 5-(4-Methoxy-3-methylphenyl)-5-oxovalerate

  • MDL number MFCD09801996
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7739C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7739C
Availability details for
Quantity
  • Availability
  • Add to Cart