RIE156331132 Rieke Metals, Inc.

Ethyl 4-(2-chlorophenyl)-4-oxobutyrate

  • MDL number MFCD03844216
  • Cas Number : 898759-01-8
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7846B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7846B
Availability details for
Quantity
  • Availability
  • Add to Cart