RIE156331136 Rieke Metals, Inc.

ethyl 8-(2-chlorophenyl)-8-oxooctanoate

  • MDL number MFCD03844220
  • Cas Number : 898759-09-6
  • Molecular Weight : 296.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7846F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7846F
Availability details for
Quantity
  • Availability
  • Add to Cart