RIE156331137 Rieke Metals, Inc.

(1,3-Dioxolan-2-ylmethyl)cyclohexane

  • MDL number MFCD07700178
  • Cas Number : 898759-11-0
  • Molecular Weight : 170.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8042A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8042A
Availability details for
Quantity
  • Availability
  • Add to Cart