RIE156331140 Rieke Metals, Inc.

1,3-Dichloro-2-(1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD04117653
  • Cas Number : 898759-17-6
  • Molecular Weight : 233.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8045A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8045A
Availability details for
Quantity
  • Availability
  • Add to Cart