RIE156331142 Rieke Metals, Inc.

3-(1,3-dioxolan-2-ylmethyl)-1,2-dimethoxybenzene

  • MDL number MFCD04117659
  • Cas Number : 898759-21-2
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8052A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8052A
Availability details for
Quantity
  • Availability
  • Add to Cart