RIE156331143 Rieke Metals, Inc.

2-(1,3-Dioxolan-2-ylmethyl)-1,4-dimethoxybenzene

  • MDL number MFCD04117661
  • Cas Number : 898759-24-5
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8054A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8054A
Availability details for
Quantity
  • Availability
  • Add to Cart