RIE156331145 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-ylmethyl)-1,3-dimethoxybenzene

  • MDL number MFCD04117663
  • Cas Number : 898759-30-3
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8056A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8056A
Availability details for
Quantity
  • Availability
  • Add to Cart