RIE156331148 Rieke Metals, Inc.

3,4-Dimethyl-1-(1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD04117666
  • Cas Number : 898759-39-2
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8059A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8059A
Availability details for
Quantity
  • Availability
  • Add to Cart