RIE156331150 Rieke Metals, Inc.

2-(1,3-dioxolan-2-ylmethyl)-1-ethoxybenzene

  • MDL number MFCD04117668
  • Cas Number : 898759-45-0
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8062A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8062A
Availability details for
Quantity
  • Availability
  • Add to Cart