RIE156331153 Rieke Metals, Inc.

1-(1,3-Dioxolan-2-ylmethyl)-3-methylbenzene

  • MDL number MFCD04117678
  • Cas Number : 898759-51-8
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8082A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8082A
Availability details for
Quantity
  • Availability
  • Add to Cart