RIE156331154 Rieke Metals, Inc.

4-(1,3-Dioxolan-2-ylmethyl)biphenyl

  • MDL number MFCD04117679
  • Cas Number : 898759-54-1
  • Molecular Weight : 240.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8090A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8090A
Availability details for
Quantity
  • Availability
  • Add to Cart