RIE156331157 Rieke Metals, Inc.

4-Fluoro-3-methyl (1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD07700179
  • Cas Number : 898785-07-4
  • Molecular Weight : 196.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8100A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8100A
Availability details for
Quantity
  • Availability
  • Add to Cart