RIE156331161 Rieke Metals, Inc.

3-Fluoro-4-methoxy (1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD08064178
  • Cas Number : 898785-19-8
  • Molecular Weight : 212.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8104A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8104A
Availability details for
Quantity
  • Availability
  • Add to Cart