RIE156331162 Rieke Metals, Inc.

5-Fluoro-2-methoxy (1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD08064179
  • Cas Number : 898785-22-3
  • Molecular Weight : 212.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8105A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8105A
Availability details for
Quantity
  • Availability
  • Add to Cart