RIE156331164 Rieke Metals, Inc.

4-methoxy-3-methyl(1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD08064181
  • Cas Number : 898785-28-9
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8107A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8107A
Availability details for
Quantity
  • Availability
  • Add to Cart