RIE156331165 Rieke Metals, Inc.

2-methoxy-5-methyl(1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD08064182
  • Cas Number : 898785-31-4
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8108A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8108A
Availability details for
Quantity
  • Availability
  • Add to Cart