RIE156331166 Rieke Metals, Inc.

2,4,6-Trimethyl(1,3-dioxolan-2-ylmethyl)benzene

  • MDL number MFCD08064183
  • Cas Number : 898785-34-7
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8109A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8109A
Availability details for
Quantity
  • Availability
  • Add to Cart