RIE156331179 Rieke Metals, Inc.

4-(5,5-dimethyl-1,3-dioxan-2-yl)-2'-iodobutyrophenone

  • MDL number MFCD03844223
  • Cas Number : 898785-54-1
  • Molecular Weight : 388.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8514-3-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8514-3-1
Availability details for
Quantity
  • Availability
  • Add to Cart