RIE156331184 Rieke Metals, Inc.

5-(5,5-dimethyl-1,3-dioxan-2-yl)-4'-iodovalerophenone

  • MDL number MFCD03844228
  • Cas Number : 898785-64-3
  • Molecular Weight : 402.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8514-4-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8514-4-3
Availability details for
Quantity
  • Availability
  • Add to Cart