RIE156331189 Rieke Metals, Inc.

4'-bromo-4-(5,5-dimethyl-1,3-dioxan-2-yl)butyrophenone

  • MDL number MFCD03844231
  • Cas Number : 898785-74-5
  • Molecular Weight : 341.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8515-3-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8515-3-3
Availability details for
Quantity
  • Availability
  • Add to Cart