RIE156331199 Rieke Metals, Inc.

3'-chloro-5-(5,5-dimethyl-1,3-dioxan-2-yl)valerophenone

  • MDL number MFCD03844239
  • Cas Number : 898785-94-9
  • Molecular Weight : 310.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8516-4-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8516-4-2
Availability details for
Quantity
  • Availability
  • Add to Cart