RIE156331202 Rieke Metals, Inc.

3-(1,3-Dioxan-2-yl)-3'-fluoropropiophenone

  • MDL number MFCD02261798
  • Cas Number : 898786-00-0
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8517-2-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8517-2-2
Availability details for
Quantity
  • Availability
  • Add to Cart