RIE156331203 Rieke Metals, Inc.

3-(1,3-dioxan-2-yl)-4'-fluoropropiophenone

  • MDL number MFCD02261799
  • Cas Number : 898786-02-2
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8517-2-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8517-2-3
Availability details for
Quantity
  • Availability
  • Add to Cart