RIE156331205 Rieke Metals, Inc.

4-(5,5-dimethyl-1,3-dioxan-2-yl)-3'-fluorobutyrophenone

  • MDL number MFCD03844242
  • Cas Number : 898786-06-6
  • Molecular Weight : 280.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8517-3-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8517-3-2
Availability details for
Quantity
  • Availability
  • Add to Cart