RIE156331208 Rieke Metals, Inc.

5-(5,5-dimethyl-1,3-dioxan-2-yl)-3'-fluorovalerophenone

  • MDL number MFCD03844245
  • Cas Number : 898786-15-7
  • Molecular Weight : 294.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8517-4-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8517-4-2
Availability details for
Quantity
  • Availability
  • Add to Cart