RIE156331209 Rieke Metals, Inc.

5-(5,5-dimethyl-1,3-dioxan-2-yl)-4'-fluorovalerophenone

  • MDL number MFCD03844246
  • Cas Number : 898786-18-0
  • Molecular Weight : 294.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8517-4-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8517-4-3
Availability details for
Quantity
  • Availability
  • Add to Cart