RIE156331210 Rieke Metals, Inc.

3-(1,3-dioxan-2-yl)-3'-nitropropiophenone

  • MDL number MFCD02261801
  • Cas Number : 898786-21-5
  • Molecular Weight : 265.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8518-2-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8518-2-2
Availability details for
Quantity
  • Availability
  • Add to Cart