RIE156331214 Rieke Metals, Inc.

4-(5,5-dimethyl-1,3-dioxan-2-yl)-3'-nitrovalerophenone

  • MDL number MFCD03844249
  • Cas Number : 898786-33-9
  • Molecular Weight : 321.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8518-4-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8518-4-2
Availability details for
Quantity
  • Availability
  • Add to Cart