RIE156331218 Rieke Metals, Inc.

3-(1,3-dioxan-2-yl)-4'-methoxypropiophenone

  • MDL number MFCD02261805
  • Cas Number : 121789-38-6
  • Molecular Weight : 250.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8519-2-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8519-2-3
Availability details for
Quantity
  • Availability
  • Add to Cart