RIE156331222 Rieke Metals, Inc.

5-(5,5-dimethyl-1,3-dioxan-2-yl)-2'-methoxyvalerophenone

  • MDL number MFCD03844254
  • Cas Number : 898786-48-6
  • Molecular Weight : 306.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8519-4-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8519-4-1
Availability details for
Quantity
  • Availability
  • Add to Cart