RIE156331223 Rieke Metals, Inc.

5-(5,5-dimethyl-1,3-dioxan-2-yl)-3'-methoxyvalerophenone

  • MDL number MFCD03844255
  • Cas Number : 898786-51-1
  • Molecular Weight : 306.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8519-4-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8519-4-2
Availability details for
Quantity
  • Availability
  • Add to Cart