RIE156331234 Rieke Metals, Inc.

3',5'-dimethyl-4'-methoxy-3-(1,3-dioxan-2-yl)propiophenone

  • MDL number MFCD02261809
  • Cas Number : 898786-73-7
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8521-2-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8521-2-0
Availability details for
Quantity
  • Availability
  • Add to Cart