RIE156331240 Rieke Metals, Inc.

2',4'-difluoro-5-(5,5-dimethyl-1,3-dioxan-2-yl)valerophenone

  • MDL number MFCD03844268
  • Cas Number : 898786-85-1
  • Molecular Weight : 312.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8523-4-0

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8523-4-0
Availability details for
Quantity
  • Availability
  • Add to Cart