RIE156331249 Rieke Metals, Inc.

3-(1,3-dioxan-2-yl)-4'-ethylpropiophenone

  • MDL number MFCD02261816
  • Cas Number : 898787-08-1
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8528-2-3A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8528-2-3A
Availability details for
Quantity
  • Availability
  • Add to Cart