RIE156331250 Rieke Metals, Inc.

4'-n-butyl-3-(1,3-dioxan-2-yl)propiophenone

  • MDL number MFCD02261818
  • Cas Number : 898787-13-8
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8528-2-3C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8528-2-3C
Availability details for
Quantity
  • Availability
  • Add to Cart