RIE156331251 Rieke Metals, Inc.

3-(1,3-Dioxan-2-yl)-4'-hexylpropiophenone

  • MDL number MFCD02261820
  • Cas Number : 898787-17-2
  • Molecular Weight : 304.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/8528-2-3E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/8528-2-3E
Availability details for
Quantity
  • Availability
  • Add to Cart